Anti-GATA3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 16E10A23]

Supplier: Biolegend
653807 653808
10763-466EA 165.5 USD
10763-466 10763-468
Anti-GATA3 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 16E10A23]
Antibodies
Alexa Fluor® 488 anti-GATA3 [16E10A23]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GATA3
Clonality: Monoclonal
Clone: 16E10A23
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol GATA3
Environmentally Preferable
Antigen Synonyms trans-acting T-cell-specific transcription factor GATA-3|GATA-binding factor 3
Antibody Type Primary
Clonality Monoclonal
Conjugation Alexa Fluor® 488
Clone 16E10A23
Reactivity Human,Mouse
Host Mouse
Gene ID 2625, 14462
Isotype IgG2b kappa
Immunogen Partial human GATA3 recombinant protein (1-258 aa).
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 488 under optimal conditions.
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR