Hydrogen Peroxide WFI Sterile Solutions, 6%
Supplier: Steris
1S0477 1S0496 1S0408 1S3215WR
10755-294CS 811.2 USD
10755-294 10755-296 10755-298 76423-316
Hydrogen Peroxide WFI Sterile Solutions, 6%
Cleaning Agents Detergents and Surface Cleansers
STERIS' 6% Hydrogen Peroxide WFI sterile solution removes residue from hard surfaces even from the most critical surfaces. It is formulated to be a low residue cleaner with minimal organic carbon.
  • Quality - Extremely low level of endotoxins from WFI produced by a Finn-Aqua® Multiple Effect Water Still
  • Ease of Use - Ready to use sterile hydrogen peroxide solution with extremely low odor and residue
  • Convenience - Double bagged for easy introduction into your aseptic areas
  • Safety - 0.2 µm filtered, aseptically filled and packaged minimizes potential for
More Product Information