Stainless Steel Quick Disconnect Handles, Micronova
Supplier: Micronova
SSU-1 SSU-3 SSU-6 SSU-7 SSU-9 SSU-8 QDAH-13 QDAH-19 SSU-2 SSU-4 SSU-5 QDAH-08
68400-164EA 294.11 USD
68400-164 68400-166 68400-302 68400-304 89040-932 89040-934 94023-796 94023-798 68400-163 68400-522 89428-202 10753-438
Stainless Steel Quick Disconnect Handles, Micronova
Mops
Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.
  • Universal handles work with all Quick Disconnect tools and mop adapters
  • 304 electropolished stainless steel with polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available in a range of fixed or extendable lengths from 8 in (20 cm) to 10 ft (3 m)
  • Quick Disconnect end attaches directly to QD mops, tools and adapters
  • SSA-3 adapter
More Product Information