Anti-UBE2S Mouse Monoclonal Antibody [clone: AT2C12]

Supplier: Abnova
MAB7885
10743-634EA 406.15 USD
10743-634
Anti-UBE2S Mouse Monoclonal Antibody [clone: AT2C12]
Antibodies
Mouse monoclonal antibody raised against full length recombinant UBE2S.

Type: Primary
Antigen: UBE2S
Clonality: Monoclonal
Clone: AT2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR