Anti-FABP4 Mouse Monoclonal Antibody [clone: 9B8D]

Supplier: Abnova
MAB10425
10712-928EA 527.69 USD
10712-928
Anti-FABP4 Mouse Monoclonal Antibody [clone: 9B8D]
Antibodies
Mouse monoclonal antibody raised against recombinant FABP4.

Recommended dilutions: Western blot: 1:500 to 1:2000; ELISA: 1:10000

Type: Primary
Antigen: FABP4
Clonality: Monoclonal
Clone: 9B8D
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR