Anti-DSG1 Mouse Monoclonal Antibody [clone: CA72/733]

Supplier: Abnova
MAB12147
10711-306EA 527.69 USD
10711-306
Anti-DSG1 Mouse Monoclonal Antibody [clone: CA72/733]
Antibodies
Mouse monoclonal antibody raised against recombinant human DSG1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: DSG1
Clonality: Monoclonal
Clone: CA72/733
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

SPECIFICATIONS

Antigen Symbol DSG1
Antigen Name Desmoglein 1
Antigen Synonyms EPKHE|SPPK1|DG1|PPKS1|EPKHIA|CDHF4|DSG
Antibody Type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone CA72/733
Host Mouse
Gene ID 1828
Isotype IgG1 kappa
Western Blot Yes
ImmunoChemistry Yes
Immunogen Recombinant protein corresponding to human DSG1.
Format Liquid
Purification Protein A/G purification
Cross Adsorption No
Storage Buffer PBS (0.05% BSA, 0.05% sodium azide).
Storage Temperature Store at 4°C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR