Anti-UBR2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-9144R-FITC
10667-940EA 803.92 USD
10667-940
Anti-UBR2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). Ubr2 (Ubiquitin-protein ligase E3-alpha-2), also known as N-recognin-2, is a 1755 amino acid protein that contains one UBR-type zinc finger and one RING-type zinc finger. Participating in protein modification events within the N-end rule pathway, Ubr2 functionsMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol UBR2
Antigen Name Ubiquitin Protein Ligase E3 Component N-recognin 2
Antigen Synonyms C6orf133|Ubiquitin-protein ligase E3-alpha-2|N-recognin-2|Ubiquitin-protein ligase E3-alpha-II|E3 ubiquitin-protein ligase UBR2|KIAA0349
Antibody Type Primary
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 23304
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross Adsorption No
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at –20 °C for 12 months
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR