Anti-ASB13 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-9106R-HRP
10667-212EA 803.92 USD
10667-212
Anti-ASB13 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the Elongin B/C complex. Several other families of proteins also contain SOCS boxes, but differ from the SOCS proteins in the type of domain they contain upstream of the SOCS box. The largest family of SOCS box-containing proteins is the ankyrin repeat and SOCS box-containing (ASB) protein family. ASB-13 (ankyrin repeat and SOCS box-containing 13) is a 278 amino acid member of the ASB familyMore Product Information