Anti-TEK Mouse Monoclonal Antibody [clone: 3C1]

Supplier: Abnova
H00007010-M06
10603-550EA 335 USD
10603-550
Anti-TEK Mouse Monoclonal Antibody [clone: 3C1]
Antibodies
Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: 3C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR