E. coli host Cell protein ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT127-0001
10551-284EA 1034.41 USD
10551-284
E. coli host Cell protein ELISA Kit, Enzo Life Sciences
Assays ELISAs
Sensitive (30 ng/ml) ELISA for the quantitative determination of host cell protein contamination in bulk products expressed in E
For the quantitative determination of contamination with E.coli host cell protein.
The E. coli host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of E. coli host cell protein contamination with results in 3 hours
coli expression systems.
Información para pedidos: Contains 1 x 96 well plate, protein standards, 5x dilution buffer, 10x PBS-T, reporting antibody, streptavidin-HRP conjugate, TMB substrate, stop solution, plate sealer.