Anti-Dihydropyrimidinase Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8287R
10481-136EA 631.29 USD
10481-136
Anti-Dihydropyrimidinase Rabbit Polyclonal Antibody
Antibodies
Catalyzes the second step of the reductive pyrimidine degradation, the reversible hydrolytic ring opening of dihydropyrimidines. Can catalyzes the ring opening of 5,6-dihydrouracil to N-carbamyl-alanine and of 5,6-dihydrothymine to N-carbamyl-amino isobutyrate.Tissue specificity:Liver and kidney.Involvement in disease:Defects in DPYS are the cause of dihydropyrimidinase deficiency (DHPD). DHPD is an autosomal recessive disorder characterized by dihydropyrimidinuria and associated with a variable clinical phenotype: epileptic or convulsive attacks, dysmorphic features and severe developmentalMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol Dihydropyrimidinase
Antigen Name Dihydropyrimidinase
Antigen Synonyms DHPase|Dpys|Dihydropyrimidine amidohydrolase|DPYS_HUMAN|Dihydropyrimidinase|Hydantoinase.|DHP
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 1807
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross Adsorption No
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage Temperature Store at –20 °C for 12 months
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR