Anti-COLEC10 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-8033R
10474-906EA 631.29 USD
10474-906
Anti-COLEC10 Rabbit Polyclonal Antibody
Antibodies
Lectin that binds to various sugars: galactose >mannose = fucose >N-acetylglucosamine >N-acetylgalactosamine.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: COLEC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR