Anti-RACGAP1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-7766R-CY3
10469-326EA 803.92 USD
10469-326
Anti-RACGAP1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Rho GTPases control a variety of cellular processes. There are 3 subtypes of Rho GTPases in the Ras superfamily of small G proteins: RHO, RAC and CDC42. GTPase-activating proteins (GAPs) bind activated forms of Rho GTPases and stimulate GTP hydrolysis. Through this catalytic function, Rho GAPs negatively regulate Rho-mediated signals. GAPs may also serve as effector molecules and play a role in signaling downstream of Rho and other Ras-like GTPases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RACGAP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR