Anti-RAB9A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-6177R
10432-208EA 631.09 USD
10432-208
Anti-RAB9A Rabbit Polyclonal Antibody
Antibodies
RAB proteins are GTPases that regulate vesicular trafficking and reside in specific intracellular compartments. RAB9 has been localized to components of the endocytic/exocytic pathway. It has been implicated in the recycling of membrane receptors, such as the mannose 6-phosphate receptor from early endosomes to the trans Golgi network.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAB9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR