Anti-APEH Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5983R-CY3
10427-820EA 803.92 USD
10427-820
Anti-APEH Rabbit Polyclonal Antibody (Cy3®)
Antibodies
AARE (Acylamino-acid-releasing enzyme) is also known as Acyl-peptide hydrolase. It catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. It can play an important role in destroying oxidatively damaged proteins in living cells. Deletions of this gene locus corresponding to the protein are found in various types of carcinomas, including small cell lung carcinoma and renal cell carcinoma.

Recommended Dilutions: IF(IHC-More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol APEH
Antigen Name Acylaminoacyl-Peptide Hydrolase
Antigen Synonyms D3F15S2|Acylamino-acid-releasing enzyme|Acyl-peptide hydrolase|APEH|AARE|D3S48E|Acylaminoacyl-peptidase|APH|DNF15S2|Oxidized protein hydrolase|OPH
Antibody Type Primary
Clonality Polyclonal
Conjugation Cy3®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 327
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross Adsorption No
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR