Human Recombinant APRIL (from HEK293), Unconjugated

Supplier: ACROBIOSYSTEMS INC MS

APLH52561MG
103888-822EA 2487.69 USD
103888-822 103888-824
Human Recombinant APRIL (from HEK293), Unconjugated
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/Peptide Name APRIL
Protein Synonyms APRIL,TNFSF13,TALL-2,TRDL-1,CD256,TALL2,ZTNF2
Environmentally Preferable
Species Human
Source
Conjugation Unconjugated
Gene ID O75888-1;8741;NP_003799.1
Endotoxin Level Low
Endotoxin Content Less than 0.1 EU per μg by the LAL method.
Biological Activity Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Purity >95% as determined by SDS-PAGE.
Formulation Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Molecular Weight 43.2 kDa
Storage Conditions –20 °C
Shipping Temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR