Anti-PRKCG Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-3730R-CY5.5
10386-998EA 823.29 USD
10386-998
Anti-PRKCG Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. There have been at least 12 different PKCMore Product Information