Anti-Bevacizumab Hybridoma Polyclonal Antibody (For Neutralizing assay)

Supplier: ACROBIOSYSTEMS INC MS
BEBY121MG BEBY12100UG
103815-438EA 2417.69 USD
103815-438 103815-440
Anti-Bevacizumab Hybridoma Polyclonal Antibody (For Neutralizing assay)
Antibodies
Type: Primary
Antigen: Bevacizumab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR