T4 DNA Polymerase
Supplier: INTACT GENOMICS, INC MS
3222
103753-186EA 201.92 USD
103753-186
T4 DNA Polymerase
Enzymes
T4 DNA polymerase which catalyzes DNA synthesis in the 5' - 3' direction and requires the presence of template and primer.
  • Useful for probe labeling using replacement synthesis, DNA library preparation for next-generation sequencing, and more
  • Extremely high purity
  • Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
  • The physical purity of this enzyme is ≥95% as assessed by SDS-PAGE with Coomassie® blue staining
  • 10X T4 DNA polymerase buffer
T4 DNA polymerase has both a DNA-dependent DNA polymerase activity and a potent 3' - 5' exonuclease activity.
Información de suministro: All our products are shipped on dry ice.