Chromatography Columns, Pre-packed, Cytiva
Supplier: Cytiva
29321909 29315187 29340185 29321910 29315186 29363635 29315195
103743-734EA 2797.38 USD
103743-734 103743-736 103743-738 103743-744 103743-746 103743-748 103743-750
Chromatography Columns, Pre-packed, Cytiva
Chromatography Columns
Prepacked validation columns are suitable for scale-down studies such as viral clearance.
  • High-quality packing
  • Excellent dimensions for scale-down studies
  • Prepacked with the most commonly used BioProcess resins
  • High consistency between columns in terms of efficiency and asymmetry
Results of Analysis certificate available for each individual column - includes column performance testing such as asymmetry and efficiency. Developed together with biopharmaceutical experts to ensure optimal function for scale-down studies such as viral clearance.