Influenza A H1N1 (A/Texas/05/2009) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
40006-V08H-100
103663-466EA 351.48 USD
103663-466
Influenza A H1N1 (A/Texas/05/2009) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of the influenza A H1N1 (A/Texas/05/2009 (H1N1)) hemagglutinin (ACP41934.1) (Met 1-Gln 529, HA1+HA2, uncleved), was fused with a C-terminal polyhistidine tag.