Influenza A H3N2 (A/Wisconsin/67/2005) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11972-V08H1-100
103633-894EA 351.48 USD
103633-894
Influenza A H3N2 (A/Wisconsin/67/2005) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the N-terminal segment (Met1-Arg345) of the influenza A hemagglutinin (A/Wisconsin/67/2005 (H3N2)) (ACF54576.1), termed as HA1, was expressed, fused with a C-terminal polyhistidine tag.