Human Recombinant UBE2I Protein (from E. coli)
Supplier: Sino Biological
13205-HNCE-100
103613-252EA 83.85 USD
103613-252 103624-362 103624-364
Human Recombinant UBE2I Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human UBE2I (P63279) (Met 1-Ser 158) was expressed and purified, with additional two amino acids (Gly and Pro) at the N-terminus.