Anti-PRL Rabbit Polyclonal Antibody

Supplier: Sino Biological
50367-T16-100
103589-068EA 142.96 USD
103589-068 103594-808 103594-810
Anti-PRL Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse PRL / Prolactin (rM PRL / Prolactin; Catalog#50367-M08B; NP_035294.2; Leu32-Cys228). PRL / Prolactin specific IgG was purified by Mouse PRL / Prolactin affinity chromatography.

Type: Primary
Antigen: PRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR