Human Hepatic Sinusoidal Endothelial Cells (HHSEC), iXCells Biotechnologies
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
10HU-021
103521-204EA 1260.31 USD
103521-204
Human Hepatic Sinusoidal Endothelial Cells (HHSEC), iXCells Biotechnologies
Cells
Human Hepatic Sinusoidal Endothelial Cells (HHSEC); Frozen; 0.5 million cells/vial
  • High-quality primary Human Hepatic Sinusoidal Endothelial Cells
  • High purity
  • IRB-approved
HHSEC are isolated from human liver. HHSEC are characterized by CD31 and vWF/Factor VIII staining. HHSEC are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.