Anti-Adenosine A2a R Mouse Monoclonal Antibody (DyLight 350) [clone: 7F6-G5-A2]

Supplier: Novus Biologicals
NBP1-39474UV
103411-628EA 712.62 USD
103411-628
Anti-Adenosine A2a R Mouse Monoclonal Antibody (DyLight 350) [clone: 7F6-G5-A2]
Antibodies
The Adenosine A2a R Antibody (7F6-G5-A2) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to Adenosine A2a R. This antibody reacts with human, mouse, rat, porcine, canine, guinea pig, rabbit. The Adenosine A2a R Antibody (7F6-G5-A2) [DyLight 350] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Adenosine A2a R
Clonality: Monoclonal
Clone: 7F6-G5-A2
Conjugation: DyLight 350
Epitope: Epitope mapped to SQPLPGER in the thirdMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR