Anti-SLC2A12 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2540R-CY5
10341-256EA 823.27 USD
10341-256
Anti-SLC2A12 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
GLUT12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. Thirteen different types of glucose/fructose transport carrier proteins designated as Glut 1-13 facilitate glucose/fructose transport across the cell membrane. Individual members of the Glut family have predicted secondary structure characteristic of 12 membrane spanning domains of other transport carriers.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC2A12
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol SLC2A12
Environmentally Preferable
Antigen Name Solute Carrier Family 2 Member A12
Antigen Synonyms GLUT8|GLUT12
Antibody Type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 154091
Isotype IgG
ImmunoChemistry Yes
Flow Cytometry Yes
Immunogen 299-317/617
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross Adsorption No
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR