Urine Preservation and Collection Devices and Kits, Precision Medical Devices
Supplier: Precision Medical Devices
124499 124500 120624 120120 124605X
103388-946CS 322.72 USD
103388-946 103388-948 103388-950 103388-968 103388-959
Urine Preservation and Collection Devices and Kits, Precision Medical Devices
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Urine preservation and collection device kits for urinalysis culture and sensitivity
  • Tubes available with or without stabilizer (preservative)
  • Multiple kit configurations available
  • Urine straw device will work with PMD urinalysis tubes, or other branded vacuum tubes
  • Preservatives include boric acid
These kits are designed for urinalysis culture and sensitivity. Use for collection, preservation, and transportation of urine. With several configurations, including a closed system to test for culture and sensitivity, PMD offers tubes with and withoutMore Product Information