TRUBLU Urine and Serum Pregnancy Test Kits, Precision Medical Devices
Supplier: Precision Medical Devices
4236725
103388-942BX 259.04 USD
103388-942
TRUBLU Urine and Serum Pregnancy Test Kits, Precision Medical Devices
Clinical Diagnostic Supplies Clinical Reagents and Kits
TRUBLU Urine and Serum hCG Pregnancy Test Kits are capable of detecting pregnancy as early as the first day of the first missed menses.
  • Sensitivity of 25 mlU/m
  • Results in three minutes for urine and five minutes for serum
  • CLIA waived for urine testing
  • Made in the USA
The TRUBLU Urine and Serum hCG Pregnancy test kit has a 25 mIU/m sensitivity, and is capable of detecting pregnancy as a early as one day after missed menses. With rapid results, in three minutes for urine and five minutes for serum, this kit offers quality and dependability.