Forensic Field Sampler, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76480-31 76480-34 76480-32 76480-33 76480-35 76480-36
103306-162KT 280.17 USD
103306-162 103306-164 103306-168 103306-170 103306-184 103306-186
Forensic Field Sampler, Electron Microscopy Sciences
Samplers Solid Samplers
Collect forensic evidence with minimum interference and/or contamination from the sampler.
  • Gunshot residue (GSR)
  • Powder and fiber samples
  • Paint chips
  • Particle sampling
  • Glass fragments
Glass specimen vial (25 OD x 55 mm high) with a high-purity certified 12.7 mm SEM pin stub specimen mount, both with and without an adhesive carbon conductive tab, mounted into the plastic cap
Choice of standard 12.7 mm SEM pin mount with 8 mm pin or 12.7 mm SEM pin mount with shorter 6 mm pin (ZEISS/LEO). Ideal for SEM specimen preparation on the most frequently usedMore Product Information