SEM Bulk Specimen Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75951-06 75951-07 75951-08 75951-09
103303-272EA 153.15 USD
103303-272 103304-802 103305-562 103306-140
SEM Bulk Specimen Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
For clamping large irregular, bulky samples to view in a scanning electron microscope.
  • Made of machined aluminum with stainless steel allen set screws
  • Pin Diameter: 3.2 mm
Información de suministro: Allen wrench included.