Anti-TCP1-beta Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88146
103271-540EA 777.05 USD
103271-540
Anti-TCP1-beta Rabbit Polyclonal Antibody
Antibodies
The TCP1-beta Antibody from Novus Biologicals is a rabbit polyclonal antibody to TCP1-beta. This antibody reacts with human, mouse, rat. The TCP1-beta Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TCP1-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR