NEBNext® Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48), New England Biolabs
Supplier: New England Biolabs (NEB)
E7560S
103258-562EA 5569 USD
103258-562
NEBNext® Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1 to 48) contains the adaptors, primers, enzymes and buffers required to convert small RNAs into indexed libraries for next-generation sequencing on the Illumina platform. The novel workflow has been optimized to minimize adaptor-dimers, while producing high-yield, high-diversity libraries.
  • Minimized adaptor-dimer contamination
  • High yields
  • Input RNA can be Total RNA
  • Suitable for methylated small RNAs (e.g., piRNAs) as well as unmethylated small RNAs