Anti-C9ORF16 Rabbit Polyclonal Antibody (FITC)
Supplier: Bioss
BS-15316R-FITC
10318-320EA 804.08 USD
10318-320
Anti-C9ORF16 Rabbit Polyclonal Antibody (FITC)
Antibodies
C9orf16 (chromosome 9 open reading frame 16) is an 83 amino acid protein that belongs to the UPF0184 (EST00098) family and is encoded by a gene that maps to human chromosome 9q34.11. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, is associatedMore Product Information