Anti-HERC1 Rabbit Polyclonal Antibody (HRP)

Supplier: Bioss

BS-15459R-HRP
10318-148EA 804.08 USD
10318-148
Anti-HERC1 Rabbit Polyclonal Antibody (HRP)
Antibodies
HERC1 is a stimulates guanine nucleotide exchange on ARF1 and Rab proteins. This protein may be involved in membrane transport processes.

Type: Primary
Antigen: HERC1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 401-500/4861
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR