Thiourea resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 3.4-3.6 mmol/g

Supplier: Supra Sciences

Synonyms: Thiocarbamide, Sulphourea

SPMP07-25 SPMP07-100 SPMP07-1KG
103061-930EA 173.85 USD
103061-930 103061-932 103061-934
Thiourea resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 3.4-3.6 mmol/g
Thiourea

Versatile scavenger of organometallic species.


Thiourea Resin, MP, is a macroporous polystyrene resin functionalized with a thiourea end group. Silica Thiourea is a silica based alkyl thioruea. Both of these reagents are exceptional scavengers of organometallic reagents with the ability to sequester Pd(0) being particularly noteworthy. MP/silica thiourea is also extremely efficient in its ability to scavenge other transition metals such as Pt, Ru, V, Ag and Hg. It has also found use as a reagent for hydrogenolysis of endoperoxides as well as functionality as a catalyst once complexed to a transition metal.

Formula: NH₂CSNH₂
MW: 76.12 g/mol
MDL Number: MFCD00008067
CAS Number: 62-56-6
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b1d4f6c2035bfa97eae0290c820b6f2dfa63ccb&vid=a83862392f98e4a6eaeeb02bd14408ebb1007391&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour Off-white
Form Granule
Loading 3.4-3.6 mmol/g
Particle size 330-1225 μm (15-50 mesh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR