Carboxylic acid resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh (>90% within) Loading: 3.4-3.6 mmol/g
Supplier: Supra Sciences
SPMP29-100 SPMP29-1KG
103061-878EA 98 USD
103061-878 103061-880
Carboxylic acid resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh (>90% within) Loading: 3.4-3.6 mmol/g
Carboxylic acid resin
Solid supported carboxylic acid utilized in solid phase synthesis for coupling amino acids at amino terminus.
Carboxylic Acid Resin, MP, is a macroporous resin functionalized with a carboxylic acid acid end group. It is a weak cation exchange resin, and has a different scavenging profile than MP-TsOH (SCX) due to its weaker acidity. The resin is designed to perform well in affinity chromatography based separation of peptides and proteins under aqueous elution conditions.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9177a879fd054fbd1317903d68502359302eceee&vid=1487892a8f094ce42661418efc2ea09c0c789ea0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Colour Off-white
Form Granule
Loading 3.4-3.6 mmol/g
Particle size 330-1225 μm (15-50 mesh)