Human Recombinant CD46 (from HEK293 Cells)

Supplier: ACROBIOSYSTEMS INC MS

CD6H522650UG
103014-096EA 350 USD
103014-096
Human Recombinant CD46 (from HEK293 Cells)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/Peptide Name CD46
Environmentally Preferable
Protein Synonyms CD46|MIC10|MCP|TRA2.10|AHUS2|TLX
Protein/Peptide Type Recombinant
Protein Function Bio-Markers & CD Antigens
Species Human
Source HEK293 cells
Conjugation Unconjugated
Protease-free Y
Animal-Free Y
Endotoxin Level Low
Endotoxin Content Less than 1.0 EU per μg by the LAL method.
UniProtKB P15529
Biological Activity Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Purity >95% as determined by SDS-PAGE.
Formulation Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Molecular Weight 33.6 kDa
Storage Conditions –20 °C
Shipping Temperature Powder,RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR