Anti-GUCY1A3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-13544R-CY5
10300-330EA 823.27 USD
10300-330
Anti-GUCY1A3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase. The soluble forms, known as GCS or sGC, act as receptors for nitric oxide. The membrane-bound receptor forms, known as GC, are peptide hormone receptors. GCS, a cGMP-synthesizing enzyme, is the major receptor for the neurotransmitter nitric oxide (NO). It plays a crucial role in smooth muscle contractility, platelet reactivity and neurotransmission. GCS is a heme containing heterodimer, consisting of one alpha subunit, designated GCS-alpha-1, and one beta subunit. TheMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol GUCY1A3
Antigen Name Guanylyl Cyclase alpha 3
Antigen Synonyms MYMY6|GC-SA3|GUCSA3|GUC1A3|GUCY1A1|GUCA3
Antibody Type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 2982
Isotype IgG
ImmunoChemistry Yes
Immunogen 330-380/690
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross Adsorption No
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR