Anti-DMPO Goat Polyclonal Antibody (Ovalbumin)

Supplier: Eagle Biosciences
DMP00-A100
102994-254EA 882.69 USD
102994-254
Anti-DMPO Goat Polyclonal Antibody (Ovalbumin)
Antibodies
Type:
Antigen: DMPO
Clonality: Polyclonal
Clone:
Conjugation: Ovalbumin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR