Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)

Supplier: Li-Cor
925-32210
102971-154EA 150.12 USD
102971-154
Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using pooled mouse IgG covalently linked to agarose beads. Based on ELISA and flow cytometry, this antibody reacts with the heavy and light chains of mouse IgG1, IgG2a, IgG2b, and IgG3, and with the light chains of mouse IgM and IgA. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with goat, horse, human, rabbit, and rat serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blotMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol IgG
Environmentally Preferable
Antigen Name Immunoglobulin G
Antibody Type Secondary
Clonality Polyclonal
Conjugation IRDye® 800CW
Reactivity Mouse
Host Goat
Isotype
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Form Lyophilized
Cross Adsorption Highly Cross-adsorbed
Storage Buffer PBS, pH 7.4. Contains 10 mg/mL BSA (free of IgG and protease) as stabilizer and 0.1% sodium azide as preservative, after reconstitution
Storage Temperature Protect from light. Store at 4 °C prior to reconstitution
Tested Applications On-Cell Western Assay, Protein Array, Microscopy, 2D Gel Detection, Tissue Section Imaging, Small Animal Imaging, Virus Titration, FRET-based Assay = Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR