Anti-IgM Goat Polyclonal Antibody (IRDye® 680LT)
Supplier: Li-Cor
925-68080
102971-036EA 150.12 USD
102971-036
Anti-IgM Goat Polyclonal Antibody (IRDye® 680LT)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgM covalently linked to agarose beads. Based on ELISA and/or flow cytometry, this antibody reacts with the heavy chain of mouse IgM. This antibody was tested by Dot Blot and/or solid phase adsorbed for minimal cross-reactivity with mouse IgG1, IgG2a, IgG3, and IgA, pooled human sera and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: WesternMore Product Information