293LTV Cell Line, Cell Biolabs
Supplier: Cell Biolabs
LTV-100
102963-884EA 523.08 USD
102963-884
293LTV Cell Line, Cell Biolabs
Cells
The 293LTV Cell Line is derived from the parental 293T cells but selected for attributes that increase lentiviral production, including firmer attachment and larger surface area
This allows the cells to produce higher levels of lentivirus compared to standard 293T cells.