Human Recombinant SERPINI2/Serpin I2/Myoepithelium-derived serine protease inhibitor/Pancpin/Pancreas-specific protein TSA2004/Peptidase inhibitor 14/PI-14/MEPI/PI14/
Supplier: Bon Opus Biosciences
CJ02-10UG CJ02-50UG CJ02-500UG CJ02-1MG
102944-218EA 118.46 USD
102944-218 102944-220 102944-222 102944-224
Human Recombinant SERPINI2/Serpin I2/Myoepithelium-derived serine protease inhibitor/Pancpin/Pancreas-specific protein TSA2004/Peptidase inhibitor 14/PI-14/MEPI/PI14/
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Serpin I2 is produced by our Mammalian expression system and the target gene encoding Ser19-Leu405 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.