Human Recombinant Cyclin-dependent kinase 2-associated protein 2

Supplier: Bon Opus Biosciences

C785-10UG C785-50UG C785-500UG C785-1MG
102942-922EA 57.08 USD
102942-922 102942-924 102942-926 102942-928
Human Recombinant Cyclin-dependent kinase 2-associated protein 2
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human CDK2AP2 is produced by our Mammalian expression system and the target gene encoding Met1-Thr126 is expressed with a 6His tag at the C-terminus.


Información accesorios: Accessories not included.


Información de suministro: Shipped Via FedEx


Envase: Shipped at ambient temperature via FedEx envelope.


Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now

SPECIFICATIONS

Protein/Peptide Name Cyclin-dependent kinase 2-associated protein 2 Cyclin-dependent kinase 2-associated protein 2
Environmentally Preferable
Protein Synonyms CDK2-associated protein 2|DOC-1R|CDK2AP2|DOC-1-related protein|DOC1R CDK2-associated protein 2|DOC-1R|CDK2AP2|DOC-1-related protein|DOC1R
Protein/Peptide Type Recombinant Recombinant
Species Mouse Human
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR