4-(Oxetan-3-yloxy)aniline ≥95%

Supplier: Matrix Scientific

115938-5G
102899-914EA 6255.38 USD
102899-914
4-(Oxetan-3-yloxy)aniline ≥95%
4-(Oxetan-3-yloxy)aniline

Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD20233076
CAS Number: 1349715-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR