2-(2-Methoxyphenyl)acetyl chloride ≥95%

Supplier: Matrix Scientific

113657-1G
102897-988EA 426.15 USD
102897-988
2-(2-Methoxyphenyl)acetyl chloride ≥95%
2-(2-Methoxyphenyl)acetyl chloride

Hazard: Irritant

Formula: C₉H₉ClO₂
MDL Number: MFCD08899364
CAS Number: 28033-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR