N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium imidodisulfuryl fluoride ≥98%

Supplier: Matrix Scientific

Synonyms: Diethyl(2-methoxyethyl)methylammonium Bis(fluorosulphonyl)imide

115476-1G 115476-5G
102896-440EA 330.77 USD
102896-440 102896-442
N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium imidodisulfuryl fluoride ≥98%
N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium imidodisulfuryl fluoride

Hazard: Irritant

Formula: C₈H₂₀F₂N₂O₅S₂
MW: 326.38 g/mol
MDL Number: MFCD22421701
CAS Number: 1079129-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR