2-Azido-N-(2-ethyl-6-methylphenyl)acetamide 97

Supplier: Matrix Scientific

115044-500MG 115044-1G
102895-270EA 693.85 USD
102895-270 102895-272
2-Azido-N-(2-ethyl-6-methylphenyl)acetamide 97
2-Azido-N-(2-ethyl-6-methylphenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₄N₄O
MW: 218.26 g/mol
MDL Number: MFCD14652806
CAS Number: 1249964-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR