Methyl (2E)-pent-2-enoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl trans-2-pentenoate, (E)-Methyl pent-2-enoate

114670-1G 114670-10G 114670-50G
102894-472EA 26.15 USD
102894-472 102894-474 102894-476
Methyl (2E)-pent-2-enoate ≥97%
Methyl (2E)-pent-2-enoate

Hazard: Flammable/Store in Freezer

Formula: C₆H₁₀O₂
MW: 114.144 g/mol
Boiling Pt: 142…145 °C
Flash Pt: 16…26 °C
MDL Number: MFCD00137611
CAS Number: 15790-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR